ENA447958649 Enamine

5-chloro-4H-1,2,4-triazol-3-ol

  • MDL number MFCD09995740
  • Cas Number : 1003-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209662
Availability details for
Quantity
  • Availability
  • Add to Cart