ENA447958652 Enamine

1-[(4-iodophenyl)carbamoyl]cyclopropane-1-carboxylic acid

  • MDL number MFCD16676105
  • Cas Number : 1247901-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209665
Availability details for
Quantity
  • Availability
  • Add to Cart