ENA447958655 Enamine

5-bromo-2-(chloromethyl)-1,3-benzoxazole

  • MDL number MFCD10696614
  • Cas Number : 110704-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-65
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210052
Availability details for
Quantity
  • Availability
  • Add to Cart