ENA447958656 Enamine

2-chloro-1-(oxan-4-yl)ethan-1-one

  • MDL number MFCD23844919
  • Cas Number : 1537276-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210059
Availability details for
Quantity
  • Availability
  • Add to Cart