ENA447958658 Enamine

tert-butyl 4-(2-amino-1,3-thiazol-5-yl)piperidine-1-carboxylate

  • MDL number MFCD24467163
  • Cas Number : 820231-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210063
Availability details for
Quantity
  • Availability
  • Add to Cart