ENA447958667 Enamine

1-(3,4-dimethoxy-2-methylphenyl)propan-2-one

  • MDL number MFCD03428558
  • Cas Number : 7537-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208765
Availability details for
Quantity
  • Availability
  • Add to Cart