ENA447958669 Enamine

7-hydroxy-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD12024481
  • Cas Number : 59647-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208768
Availability details for
Quantity
  • Availability
  • Add to Cart