ENA447958672 Enamine

2-(oxolan-3-yl)-1H-1,3-benzodiazole

  • MDL number MFCD14610568
  • Cas Number : 906519-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208792
Availability details for
Quantity
  • Availability
  • Add to Cart