ENA447958675 Enamine

2,7-dichloroimidazo[1,2-a]pyridine-3-carbonitrile

  • MDL number MFCD22013301
  • Cas Number : 1368306-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210096
Availability details for
Quantity
  • Availability
  • Add to Cart