ENA447958676 Enamine

2-chloro-4-methoxy-5-methylaniline

  • MDL number MFCD20696075
  • Cas Number : 344794-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210097
Availability details for
Quantity
  • Availability
  • Add to Cart