ENA447958677 Enamine

methyl 1-(hydroxymethyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD05861652
  • Cas Number : 1001519-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210098
Availability details for
Quantity
  • Availability
  • Add to Cart