ENA447958681 Enamine

ethyl 4-sulfamoylpiperidine-1-carboxylate

  • MDL number MFCD16671874
  • Cas Number : 1249785-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210108
Availability details for
Quantity
  • Availability
  • Add to Cart