ENA447958684 Enamine

4-(difluoromethyl)pyrrolidin-2-one

  • MDL number MFCD22413075
  • Cas Number : 1803589-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210112
Availability details for
Quantity
  • Availability
  • Add to Cart