ENA447958685 Enamine

4-methylpentan-2-yl chloroformate

  • MDL number MFCD24141106
  • Cas Number : 84000-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210113
Availability details for
Quantity
  • Availability
  • Add to Cart