ENA447958690 Enamine

6-bromo-2,2-dimethyl-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD11219442
  • Cas Number : 165730-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210126
Availability details for
Quantity
  • Availability
  • Add to Cart