ENA447958694 Enamine

3-amino-6-chloro-1H-isoindol-1-one

  • MDL number MFCD20696883
  • Cas Number : 65122-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210130
Availability details for
Quantity
  • Availability
  • Add to Cart