ENA447958695 Enamine

N-methyl-2-[2-(methylamino)phenoxy]aniline

  • MDL number MFCD28047960
  • Cas Number : 76967-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210131
Availability details for
Quantity
  • Availability
  • Add to Cart