ENA447958696 Enamine

1-ethyl-3-methylpiperazine

  • MDL number MFCD11858480
  • Cas Number : 428871-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208799
Availability details for
Quantity
  • Availability
  • Add to Cart