ENA447958699 Enamine

2,4-dichloro-6,6-dimethyl-5,6,7,8-tetrahydroquinazoline

  • MDL number MFCD21291789
  • Cas Number : 1480939-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208802
Availability details for
Quantity
  • Availability
  • Add to Cart