ENA447958702 Enamine

(1R)-1-(5-bromo-2,3-dihydro-1-benzofuran-7-yl)ethan-1-ol

  • MDL number MFCD26904769
  • Cas Number : 1568011-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208804
Availability details for
Quantity
  • Availability
  • Add to Cart