ENA447958710 Enamine

thiomorpholine-4-carbonyl chloride

  • MDL number MFCD20483368
  • Cas Number : 143185-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210204
Availability details for
Quantity
  • Availability
  • Add to Cart