ENA447958712 Enamine

(cyclopropylmethyl)({[2-(trifluoromethyl)phenyl]methyl})amine

  • MDL number MFCD11140266
  • Cas Number : 1019538-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208805
Availability details for
Quantity
  • Availability
  • Add to Cart