ENA447958714 Enamine

2-(dimethyl-1,2-oxazol-4-yl)propan-2-amine

  • MDL number MFCD28397707
  • Cas Number : 1520108-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210208
Availability details for
Quantity
  • Availability
  • Add to Cart