ENA447958715 Enamine

2-(2-methoxyethoxy)pyrimidin-5-amine

  • MDL number MFCD14589343
  • Cas Number : 1251024-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210209
Availability details for
Quantity
  • Availability
  • Add to Cart