ENA447958719 Enamine

4-hydroxy-2,3,6-trimethylbenzonitrile

  • MDL number MFCD22005032
  • Cas Number : 1492086-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210213
Availability details for
Quantity
  • Availability
  • Add to Cart