ENA447958720 Enamine

5-iodo-1,3-thiazole-2-carbaldehyde

  • MDL number MFCD21728910
  • Cas Number : 1540455-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210215
Availability details for
Quantity
  • Availability
  • Add to Cart