ENA447958723 Enamine

methyl 3-(aminomethyl)azetidine-1-carboxylate

  • MDL number MFCD19214596
  • Cas Number : 859164-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210220

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210220
Availability details for
Quantity
  • Availability
  • Add to Cart