ENA447958727 Enamine

4-amino-2-hydroxy-3,5-diiodobenzoic acid

  • MDL number MFCD00085725
  • Cas Number : 74764-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210234
Availability details for
Quantity
  • Availability
  • Add to Cart