ENA447958729 Enamine

4-methylpent-4-enenitrile

  • MDL number MFCD00204043
  • Cas Number : 34998-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210248
Availability details for
Quantity
  • Availability
  • Add to Cart