ENA447958730 Enamine

(5-{[(2-methylprop-2-en-1-yl)amino]methyl}furan-2-yl)methanol

  • MDL number MFCD28397558
  • Cas Number : 1592519-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208361
Availability details for
Quantity
  • Availability
  • Add to Cart