ENA447958731 Enamine

[4-(trifluoromethyl)piperidin-3-yl]methanol hydrochloride

  • MDL number MFCD21601981
  • Cas Number : 1356339-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208810
Availability details for
Quantity
  • Availability
  • Add to Cart