ENA447958736 Enamine

1-[4-(trifluoromethyl)phenyl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD15527665
  • Cas Number : 1260740-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210257

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210257
Availability details for
Quantity
  • Availability
  • Add to Cart