ENA447958740 Enamine

2-(4-aminopiperidin-1-yl)-N,N-diethylacetamide dihydrochloride

  • MDL number MFCD28397713
  • Cas Number : 1589393-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210261
Availability details for
Quantity
  • Availability
  • Add to Cart