ENA447958744 Enamine

4-chloro-2,6-diiodophenol

  • MDL number MFCD00672959
  • Cas Number : 15459-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210270
Availability details for
Quantity
  • Availability
  • Add to Cart