ENA447958745 Enamine

1-methoxybut-2-yne

  • MDL number MFCD28397714
  • Cas Number : 2768-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210271
Availability details for
Quantity
  • Availability
  • Add to Cart