ENA447958769 Enamine

1-(3-acetylphenyl)azetidin-2-one

  • MDL number MFCD27929570
  • Cas Number : 844902-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210694
Availability details for
Quantity
  • Availability
  • Add to Cart