ENA447958776 Enamine

2-amino-5-bromo-3-nitrophenol

  • MDL number MFCD23163822
  • Cas Number : 867338-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210720
Availability details for
Quantity
  • Availability
  • Add to Cart