ENA447958779 Enamine

N-(oxan-4-yl)-3-(pyridin-3-yl)imidazo[1,2-b]pyridazin-6-amine

  • MDL number MFCD26391889
  • Cas Number : 928336-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208847
Availability details for
Quantity
  • Availability
  • Add to Cart