ENA447958781 Enamine

6-ethylpyridine-3-carbaldehyde

  • MDL number MFCD13175988
  • Cas Number : 1005327-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210742
Availability details for
Quantity
  • Availability
  • Add to Cart