ENA447958782 Enamine

4-amino-2-chloro-5-fluorophenol

  • MDL number MFCD18970213
  • Cas Number : 524955-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210744
Availability details for
Quantity
  • Availability
  • Add to Cart