ENA447958784 Enamine

2-(2-iminopyrrolidin-1-yl)acetic acid

  • MDL number MFCD19229352
  • Cas Number : 16849-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210747
Availability details for
Quantity
  • Availability
  • Add to Cart