ENA447958786 Enamine

1-[1-(pyridin-2-yl)cyclobutyl]methanamine

  • MDL number MFCD23725556
  • Cas Number : 1228994-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210750
Availability details for
Quantity
  • Availability
  • Add to Cart