ENA447958787 Enamine

3-amino-2-(4-chlorophenyl)propan-1-ol hydrochloride

  • MDL number MFCD18782993
  • Cas Number : 1280721-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210766
Availability details for
Quantity
  • Availability
  • Add to Cart