ENA447958789 Enamine

2-bromopyrimidin-5-ol

  • MDL number MFCD09953508
  • Cas Number : 1240621-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208374
Availability details for
Quantity
  • Availability
  • Add to Cart