ENA447958805 Enamine

5-ethoxy-3,3-dimethyl-5-oxopentanoic acid

  • MDL number MFCD20638833
  • Cas Number : 71885-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208851
Availability details for
Quantity
  • Availability
  • Add to Cart