ENA447958807 Enamine

1-chloro-3-phenoxypropan-2-one

  • MDL number MFCD11935034
  • Cas Number : 940-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208852
Availability details for
Quantity
  • Availability
  • Add to Cart