ENA447958811 Enamine

4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD14551345
  • Cas Number : 167548-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208855
Availability details for
Quantity
  • Availability
  • Add to Cart