ENA447958820 Enamine

3-(hydroxyimino)-2,3-dihydro-1H-isoindol-1-one

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208860
Availability details for
Quantity
  • Availability
  • Add to Cart