ENA447958829 Enamine

tert-butyl 4-(pyrrolidin-3-yl)piperazine-1-carboxylate

  • MDL number MFCD06858516
  • Cas Number : 885959-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208290
Availability details for
Quantity
  • Availability
  • Add to Cart