ENA447958834 Enamine

(2,5-difluoropyridin-4-yl)methanol

  • MDL number MFCD12756645
  • Cas Number : 1227601-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210883
Availability details for
Quantity
  • Availability
  • Add to Cart